Orders LinkContactUsLinkComponent
Storage Boxes for Vials/Bottles
Transparent, 70×140×40 mm
Catalog # KOEH2-7209
Storage Boxes for Vials/Bottles
Catalog # KOEH2-7209
Storage Boxes for Vials/Bottles
Storage Boxes for Vials/Bottles
Transparent, 70×140×40 mm
Price
11,60 €
Log in for availability
1 * 1 items
Product Details & Documents
Storage boxes are suitable for bottles and vials which is used for cooling in freezers.
  • With trim insert and tight-closing lid
  • 50 places (5×10) for vessels with 12 mm Ø
Accepts autosampler bottle (1,2 ml), micro vial (1,5 ml), conic ampoule.
Specifications
  • Colour:
    Transparent
  • W×D×H:
    70×140×40 mm