Whole milk powder, NIST-1549A
Catalog # PROONIST-1549A
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pk:5
- Description:NIST-1549A, whole milk powder
- Pk Info:5× 10 g sealed aluminised pouches
Specifications
About this item
This reference standard is intended for validation of methods for determining proximates, fatty acids, cholestrol, vitamins, elements and amino acids in whole milk powder and similar materials.
Certifications: Traceable to NIST and SRM numbers.