Orders LinkContactUsLinkComponent
Pipette stand, linear
Pipette stand, linear
Universal linear stand for holding up to 11 pipettes in an upright position, White/black, 290×250×60 mm
Catalog # 613-8706
   Low Manufacturing Impact
Supplier:   Sartorius
Pipette stand, linear
Catalog # 613-8706
Supplier:  Sartorius
   Low Manufacturing Impact
Pipette stand, linear
Pipette stand, linear
Universal linear stand for holding up to 11 pipettes in an upright position, White/black, 290×250×60 mm
Price
84,70 €
Log in for availability
1 * 1 items
Product Details & Documents
This pipette stand holds pipettes in an upright position while not in use to avoid contamination from benches and tables. Designed for all Sartorius mechanical and electronic pipettes, particularly for mLINE®, Proline® Plus and Proline® mechanical devices. Also compatible with other brands.
Specifications
  • Colour:
    White/black
  • Description:
    Universal linear stand for holding up to 11 pipettes in an upright position
  • Seq:
    59
  • W×D×H:
    290×250×60 mm
  • Cat. No.:
    613-8706
  • Environmental certification:
    Renewable energy certificate; ISO 14001
  • Environmentally Preferable:
    Low Manufacturing Impact