ContactUsLinkComponent
Organic reference standards, Tioconazole assay standard, BP
Organic reference standards, Tioconazole assay standard, BP
Certificates
Organic reference standards, Tioconazole assay standard, BP
Specifications
  • CAS No.:
    65899-73-2
  • Formula:
    C₁₆H₁₃Cl₃N₂OS
  • Description:
    Tioconazole assay standard