Organic reference standards, thiamine mononitrate assay standard, BP
PROOBP414
:
- Pk:100 mg
- Standards:Reference standard
- CAS No.:532-43-4
- Description:Thiamine mononitrate assay standard
- Formula:C₁₂H₁₇N₄OS·NO₃
- Description:Thiamine mononitrate assay standard