ContactUsLinkComponent
Microscope slides for printers
Microscope slides for printers
Catalog # KLINKP-SIL-3059
Supplier:  KLINIPATH
Microscope slides for printers
Catalog # KLINKP-SIL-3059
Supplier:  KLINIPATH
Specifications
  • Colour:
    Pink
  • Seq:
    11
  • Environmentally Preferable:
  • Pk:
    2.500
Specifications
About this item
Microscope slides for slide printers.
  • Frosted ends
  • 90° ground edges
  • Finished clipped corners of 1 mm
Allowing for easy handling of automated slide printers and digital slide imagers in slide racks.