Anti-IgE Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # 35-8027-M001
Supplier: Tonbo Biosciences
Specifications
- Antibody type:Secondary
- Antigen name:Immunoglobulin E epsilon
- Antigen symbol:IgE
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Flow cytometry:Yes
- Host:Goat
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human
- Epitope:ε chain
- Cross adsorption:Yes
- Storage temperature:Store at 2-8 °C
- Pk:1 mg
Specifications
About this item
Goat anti-Human IgE (epsilon chain) - Affinity Pure, FITC Conjugate, min x w/bovine, mouse or rabbit serum proteins
Type: Secondary
Antigen: IgE
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope: ε chain
Host: Goat
Isotype:
Reactivity: Human