Powder pillows, potassium persulphate
HACH2084766
: Hach
:
- Chemical method:Persulfate UV oxidation
- Pk:50
- Description:Powder pillows, potassium persulphate
Powder reagent in sealed package for phosphonate test and total phosphate TNT.
Powder reagent in sealed package for phosphonate test and total phosphate TNT.