Test kits, total hardness determination by titration methods
Catalog # HACH2447600
Supplier: Hach
CAS Number:
Specifications
- Pk:1
- Description:Hardness (Total) reagent set, low range
- Seq:01
- Range:0 - 2500 mg/l
Specifications
About this item
The kits are used to determine the total hardness level.
Delivery: Kit includes buffer solution (Hardness 1), ManVer® 2 hardness indicator powder pillows and TitraVer® (EDTA) and relevant titrant solution.