ContactUsLinkComponent
Cleanroom goggles, BioClean Clearview™, sterile, for single use
  111-0186
 :  Nitritex
undefined
Cleanroom goggles, BioClean Clearview™, sterile, for single use
  111-0186
 :  Nitritex
  • Type:
    Anti-fog, anti-scratch, sterile
  • Colour:
    Transparent
  • Environmentally Preferable:
  • Lenses:
    Clear PC
  • Pk:
    60
Manufactured from light weight PVC, the ultra soft single use goggles provide outstanding comfort. The specially designed indirect ventilation system allows good ventilation while reducing the incidence of particle release.
  • Toughened PC lens offering protection against flying particles
  • Lightweight frame with internal lateral vents
  • Fitted with four LLDPE indirect ventilation caps
  • Non-linting, adjustable silicon head band
Suitable for use in cleanroom class ISO 4.
 : EN 166:2001 Optical Class 1, PPE Cat II, Sterility validated to ISO 11135-1-2007
 : Double bagged in low linting Tyek/PE pouches.