Bench protectors, Scienceware®, Labmat™
Catalog # SIALL2271-1EA
Supplier: Merck
Specifications
- Packed:Roll
- Colour:White
- Description:Bench liner, Labmat™
- L×W:15200×500 mm
- Pk:1
Specifications
About this item
Disposable bench and drawer liners protect surfaces from stains and wear.
- Highly absorbent upper surface traps up to 420 ml/m² of water, dyes, corrosive chemicals or radioactive solutions
- Flexible polyethylene film backing is non-skid and prevents soak-through of most solutions
- Rolls are easily cut with a cutter or scissors while roll is retained in dispenser