dNTP set, MP Biomedicals
Supplier: MP Biomedicals
Certificates
About this item
Set consists of 100 mM aqueous solutions of dATP, dCTP, dGTP and dTTP each supplied in a separate vial.
Specifications
- Application:
- Environmentally Preferable:
Specifications
Frequently Bought Together