ContactUsLinkComponent
Cryogenic vial rack, Corning®
  734-1882
 :  Corning
   Low Manufacturing Impact
undefined
Cryogenic vial rack, Corning®
  734-1882
 :  Corning
   Low Manufacturing Impact
  • No. of Holes:
    50 (5×10)
  • Colour:
    Orange
  • Seq:
    03
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    I-RECs, ISO 14001 - Reynosa
  • Pk:
    2
This reusable PP rack holds 50 cryogenic vials.
  • Locking feature for use with all Corning® self-standing vials
  • Allows one handed opening