ContactUsLinkComponent
Pipette controller, electronic
  GLSNF161250
 :  GILSON
Pipette controller, electronic
  GLSNF161250
 :  GILSON
  • Description:
    Pipette controller, EU plug (220 V/50 Hz)
  • Pk:
    1
Pipette controller for all standard glass and plastic pipettes.
  • Ergonomic design for increased user comfort
  • Quiet motorised operation
  • Precise and smooth finger tip control of variable speed range
  • Powerful battery, continuous work for 8 hours (environmentally friendly NiMH)
  • Selectable operation modes according to volume and viscosity
  • Safety valve and filter prevent contamination
  • All parts in contact with liquid are autoclavable