- Antibody type:Primary
- Antigen name:Ring Finger Protein 8
- Antigen symbol:RNF8
- Clonality:Polyclonal
- Conjugation:Cy7®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:9025
- Antigen synonyms:E3 ubiquitin-protein ligase RNF8|hRNF8|RNF8|KIAA0646
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
The RING finger motif is a specialized DNA-binding zinc finger domain found in many transcriptional regulatory proteins. The ring finger protein (RNF) family includes any protein containing the signature RING finger motif. RNF8 is a ubiquitously expressed nuclear RING finger protein that acts as an E3 ubiquitin-protein ligase. It is required for the ubiquitination of some nuclear proteins and promotes their subsequent degradation. The heterodimeric ubiquitin-conjugating enzyme UBC13 interacts with RNF8, and they co-localize in the nucleus. RNF8 may regulate mediation of UBC13 polyubiquitylation by elongating the ubiquitin chains. RNF8 also binds to Retinoid X receptor alpha (RXR?, a member of the steroid hormone receptor superfamily. It increases RXR?mediated transactivation of the RXR?responsive element (RXRE) promoter in a dose-dependent manner, suggesting that RNF8 is a regulator of RXR?mediated transcriptional activity.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: RNF8
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat