Order Entry
Slovenia
ContactUsLinkComponent
 
Anti-FAM104B Rabbit Polyclonal Antibody (Alexa Fluor® 555)
  BOSSBS-8231R-A555
 :  Bioss
Anti-FAM104B Rabbit Polyclonal Antibody (Alexa Fluor® 555)
  BOSSBS-8231R-A555
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    Family With Sequence Similarity 104 Member B
  • Antigen symbol:
    FAM104B
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 555
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    90736
  • Antigen synonyms:
    FLJ18375|FLJ20434|CXorf44|FAM104B|member B|F104B_HUMAN|Protein FAM104B.|Family with sequence similarity 104
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at –20 °C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

 

 

The X and Y chromosomes are the human sex chromosomes. Chromosome X consists of about 153 million base pairs and nearly 1,000 genes. The combination of an X and Y chromosome lead to normal male development while two copies of X lead to normal female development. There are a number of conditions related to an unsual number and combination of sex chromosomes being inherited. More than one copy of the X chromosome with a Y chromosome causes Klinefelter's syndrome. A single copy of X alone leads to Turner's syndrome. More than 2 copies of the X chromosome, in the absence of a Y chromosome, is known as Triple X syndrome. Color blindness, hemophilia, and Duchenne muscular dystrophy are well known X chromosome-linked conditions which affect males more frequently as males carry a single X chromosome. The FAM104B gene product has been provisionally designated FAM104B pending further characterization.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: FAM104B
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human