Anti-LCMT1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # BOSSBS-6249R-FITC
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Leucine carboxyl methyltransferase 1
- Antigen symbol:LCMT1
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:51451
- Antigen synonyms:CGI-68|LCMT|Phosphatase 2A protein-leucine-carboxy methyltransferase 1|Leucine carboxyl methyltransferase 1|LCMT1|Protein-leucine O-methyltransferase|PP2A methyltransferase
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
LCMT1 catalyzes the methylation of the carboxyl group of the C-terminal leucine residue (leu309) of the catalytic subunit of protein phosphatase-2A (PPP2CA; MIM 176915)
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: LCMT1
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat