Anti-LSP1 Rabbit Polyclonal Antibody (Cy5.5®)
BOSSBS-5155R-CY5.5
: Bioss
- Antibody type:Primary
- Antigen name:Lymphocyte-specific Protein 1
- Antigen symbol:LSP1
- Clonality:Polyclonal
- Conjugation:Cy5.5®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:4046
- Antigen synonyms:WP34|pp52|F-actin binding and cytoskeleton associated protein|leukocyte-specific protein 1|lymphocyte-specific protein pp52|52 kDa phosphoprotein|leufactin (leukocyte F-actin binding protein)|lymphocyte-specific protein 1|LSP1|47 kDa actin binding protein|lymphocyte-specific antigen WP34
- Modification:phospho-Ser204
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:180-220/339
- Purification:Purified by Protein A
- Pk:100 µl
May play a role in mediating neutrophil activation and chemotaxis. This gene encodes an intracellular F-actin binding protein. The protein is expressed in lymphocytes, neutrophils, macrophages, and endothelium and may regulate neutrophil motility, adhesion to fibrinogen matrix proteins, and transendothelial migration. Alternative splicing results in multiple transcript variants encoding different isoforms. [provided by RefSeq, Jul 2008]
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: LSP1
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat