Order Entry
Slovenia
ContactUsLinkComponent
Anti-CERS2 Rabbit Polyclonal Antibody (Cy3®)
Anti-CERS2 Rabbit Polyclonal Antibody (Cy3®)
Catalog # BOSSBS-5077R-CY3
Supplier:  Bioss
Anti-CERS2 Rabbit Polyclonal Antibody (Cy3®)
Catalog # BOSSBS-5077R-CY3
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Ceramide synthase 2
  • Antigen symbol:
    CERS2
  • Clonality:
    Polyclonal
  • Conjugation:
    Cy3®
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    29956
  • Antigen synonyms:
    FLJ10243|SP260|CerS2|MGC987|LAG1 homolog ceramide Synthase 2|L3|TMSG1
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Immunogen:
    230-261/380
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

LASS2 (LAG1 homolog, ceramide synthase 2) has sequence similarity to yeast longevity assurance gene 1. It is thought to be involved in sphingolipid synthesis. Expression of LASS2 is transiently increased during the period of active myelination and is specifically localized to white matter tracts of the brain, including the Schwann cells of sciatic nerves suggesting that LASS2 is important for the synthesis of dihydroceramide used for synthesis of myelin sphingolipids. Expression of LASS2 in hepatocellular carcinoma cell lines suppresses the growth of cancer cells.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CERS2
Clonality: Polyclonal
Clone: 0
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: