Anti-BRAF Rabbit Polyclonal Antibody (Cy5.5®)
Catalog # BOSSBS-3013R-CY5.5
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Serine/threonine-protein kinase B-Raf
- Antigen symbol:BRAF
- Clonality:Polyclonal
- Conjugation:Cy5.5®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:673
- Antigen synonyms:p94|BRAF|B-Raf proto-oncogene serine/threonine-protein kinase|RAFB1|v-Raf murine sarcoma viral oncogene homolog B1|BRAF1
- Modification:phospho-Ser365
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:350-390/766
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Protein kinase involved in the transduction of mitogenic signals from the cell membrane to the nucleus. May play a role in the postsynaptic responses of hippocampal neuron. Phosphorylates MAP2K1, and thereby contributes to the MAP kinase signal transduction pathway.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: BRAF
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat