Anti-MRPL12 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
BOSSBS-3834R-FITC
: Bioss
- Antibody type:Primary
- Antigen name:Mitochondrial Ribosomal Protein L12
- Antigen symbol:MRPL12
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:6182
- Antigen synonyms:MRP-L31/34|MRPL7/L12|RPML12|mitochondrial|MRPL12|39S ribosomal protein L12|5c5-2|RM12_HUMAN.|L12mt
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
MRPL12 is a component of the 39S subunit of mammalian mitochondrial ribosomes. This protein forms homodimers whereas in prokaryotic ribosomes two L7/L12 dimers and one L10 protein form the L8 protein complex.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: MRPL12
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat