Anti-HSD11B2 Rabbit Polyclonal Antibody (Cy7®)
Catalog # BOSSBS-3618R-CY7
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Hydroxysteroid 11-beta Dehydrogenase 2
- Antigen symbol:HSD11B2
- Clonality:Polyclonal
- Conjugation:Cy7®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:3291
- Antigen synonyms:HSD11K|AME|HSD2|SDR9C3|AME1
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:170-220/405
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Catalyzes the conversion of cortisol to the inactive metabolite cortisone. Modulates intracellular glucocorticoid levels, thus protecting the nonselective mineralocorticoid receptor from occupation by glucocorticoids.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: HSD11B2
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat