Anti-URGCP Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # BOSSBS-2172R-FITC
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Upregulator of Cell Proliferation
- Antigen symbol:URGCP
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:KIAA1507|URGCP|Up-regulator of cell proliferation|HBV X protein upregulated gene 4 protein|HBxAg up-regulated gene 4 protein|URGCP_HUMAN.|HBV X protein up-regulated gene 4 protein|HBxAg upregulated gene 4 protein|Upregulated gene 4|FLJ20654|Upregulator of cell proliferation
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
URG4 may be involved in cell cycle progression through the regulation of cyclin D1 expression. It may participate in the development of hepatocellular carcinoma (HCC) by promoting hepatocellular growth and survival. URG4 may play an important role in development of gastric cancer.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: URGCP
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat