Order Entry
Slovenia
ContactUsLinkComponent
Anti-LIPG Rabbit Polyclonal Antibody (Cy7®)
Anti-LIPG Rabbit Polyclonal Antibody (Cy7®)
  BOSSBS-2397R-CY7
 :  Bioss
Anti-LIPG Rabbit Polyclonal Antibody (Cy7®)
  BOSSBS-2397R-CY7
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    Lipase endothelial
  • Antigen symbol:
    LIPG
  • Clonality:
    Polyclonal
  • Conjugation:
    Cy7®
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    9388
  • Antigen synonyms:
    EDL|EL|PRO719
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Immunogen:
    310-360/500
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

 

 

Has phospholipase and triglyceride lipase activities. Hydrolyzes high density lipoproteins (HDL) more efficiently than other lipoproteins. Binds heparin.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: LIPG
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat