Anti-CNR2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # BOSSBS-2377R-FITC
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Cannabinoid Receptor 2
- Antigen symbol:CNR2
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:1269
- Antigen synonyms:OTTHUMP00000044841|CNR3|RP11-4M23.1|testis-dominant CNR2 isoform CB2|CNR2|CB2|CX5 |HGNC:2160|cannabinoid receptor 2 (macrophage)
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:298-330/360
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Heterotrimeric G protein-coupled receptor for endocannabinoid 2-arachidonoylglycerol mediating inhibition of adenylate cyclase. May function in inflammatory response, nociceptive transmission and bone homeostasis.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: CNR2
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype:
Reactivity: