Anti-AZI1 Rabbit Polyclonal Antibody (Cy5®)
Catalog # BOSSBS-12977R-CY5
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:5-azacytidine-induced protein 1
- Antigen symbol:AZI1
- Clonality:Polyclonal
- Conjugation:Cy5®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:Azi1|Azi|RP23 37J21.1.|Pre-acalization protein 1|AZI1_HUMAN|5-azacytidine-induced protein 1|OTTMUSP00000004498|KIAA1118|Centrosomal protein 131 kDa|Centrosomal protein of 131 kDa|5 azacytidine induced 1|AZ1|Cep131|5-azacytidine induced 1
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
AZI1
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: AZI1
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat