Anti-HSP90AB1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Catalog # BOSSBS-0194R-A555
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Heat Shock Protein 90kDa Alpha (Cytosolic), Class B Member 1
- Antigen symbol:HSP90AB1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 555
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:7184
- Antigen synonyms:HSP84|HSPCB|HSPC2|Heat shock protein HSP 90-beta|HSP 90|HSP90B|Heat shock 84 kDa|HSP 84|HSP90AB1
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:550-600/803
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Molecular chaperone that functions in the processing and transport of secreted proteins. When associated with CNPY3, required for proper folding of Toll-like receptors (By similarity). Functions in endoplasmic reticulum associated degradation (ERAD). Has ATPase activity.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: HSP90AB1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat