HPLC columns, Poroshell 120 SB-C8
Supplier: Agilent
Certificates
About this item
Poroshell 120 is made with a superficially porous particle that has a solid silica core and a porous outer layer. This particle provides high efficiency at lower pressures and is ideal for fast or high-resolution separations of many types of analytes.
- Provides greater alternative selectivity
- Steric protection of the bonded phase resists degradation
- Ideal for basic, neutral, acidic samples or aggressive low pH mobile phases
The StableBond SB-8 bonded phase is made by chemically bonding a sterically-protected C8 stationary phase to the porous shell of the Poroshell 120 silica support. The densely-covered, sterically-protected diisopropyl-n-octylsilane stationary phase is chemically stable and gives longer column life.
Specifications
- Catalog No:
- HEWL821725-923
- HEWL820750-923
- HEWL823750-923
- HEWL689975-306
- HEWL683775-906
- HEWL689975-906
- HEWL683975-306
- HEWL685975-906
- HEWL689775-906
- HEWL683975-906
- HEWL685975-306
- HEWL685775-906
- Carbon load:
- 5,5%
- 5,5%
- 5,5%
- 5,5%
- 5,5%
- 5,5%
- Endcapped:
- No
- No
- No
- No
- No
- No
- Max. pressure:
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- pH range:
- 1,0 - 8,0
- 1,0 - 8,0
- 1,0 - 8,0
- 1,0 - 8,0
- 1,0 - 8,0
- 1,0 - 8,0
- Surface area:
- 130 m²/g
- 130 m²/g
- 130 m²/g
- 130 m²/g
- 130 m²/g
- 130 m²/g
- Temperature limit:
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- USP code:
- L7
- L7
- L7
- L7
- L7
- L7