J.T.Baker® BAKERBOND spe™ C18-SDB-1, SPE Columns for Special Applications
Catalog # 7650-07
Supplier: Avantor
Specifications
- Capacity:6 ml
- Description:Column with 500 mg C18 (top) and 200 mg SDB-1, separated by a PE frit
- Bed weight:700 mg
- Pk:30
Specifications
About this item
These SPE columns are designed for the extraction of pesticides from water.