Orders LinkContactUsLinkComponent
VWR® Powerpette Plus, Pipette Controller
Catalog #
VWR® Powerpette Plus, Pipette Controller
Catalog #
Supplier:  
VWR® Powerpette Plus, Pipette Controller
VWR® Powerpette Plus, Pipette Controller
Product Details & Documents
Pipette controller for glass and plastic pipettes from 1 to 100 ml.
  • Speed of suction and dispensing controlled through the finger triggers
  • Three aspirate and dispense modes
  • High and low settings both have variable speed
  • Fully charged has approximately 8 hours of continuous use available
  • Low battery light illuminates when 1 hour's power remains
Offers a unique combination of features to give precision fingertip control in a classic streamlined design, suitable for users with large or small hands. Gravity dispense mode is designed for use with 'To Deliver' (TD) pipettes.
 : Supplied with 0,45 and 0,2 µm hydrophobic filters, as standard, for effective protection against airborne contamination; 2×1,2 V nickel metal hydride batteries and universal charger.
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Multi regional charger for Powerpette Plus and Powerpette Pro
    Multi regional charger for Powerpette Plus and Powerpette Pro
    613-5849
    Log in for availability
    1 * 1 items - 20,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Replacement silicone pipette holder
    Replacement silicone pipette holder
    612-3679
    Log in for availability
    1 * 1 items
Product Information
  • Catalog #
    Availability
    Price
  • Replacement sterile filter 0,2 µm
    Replacement sterile filter 0,2 µm
    JENC266-090
    Log in for availability
    1 * 1 items - 14,10 €
Product Information
  • Catalog #
    Availability
    Price
  • Replacement filter set 0,45 µm
    Replacement filter set 0,45 µm
    612-3678
    Log in for availability
    1 * 5 items