Anti-FR-alpha Rabbit Polyclonal Antibody [clone: Y80]
Catalog # BWRLBS3861
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:FR-alpha
- Clonality:Polyclonal
- Clone:Y80
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 60-110 of Human FR-alpha.
FR-? (Y80) pAb detects endogenous levels of FR-? (Y80) protein.
Type: Primary
Antigen: FR-alpha
Clonality: Polyclonal
Clone: Y80
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat