ContactUsLinkComponent
MEM non essential amino acids 100X
MEM non essential amino acids 100X
Catalog # X0557-100
Supplier:  Biowest
CAS Number:  
undefined
MEM non essential amino acids 100X
Catalog # X0557-100
Supplier:  Biowest
CAS Number:  
Specifications
  • Pk:
    100 ml
  • Source:
  • Species:
  • Description:
    MEM NEAA, 100X, without L-glutamine (liquid)
  • Environmentally Preferable:
Specifications
About this item
Concentrated solutions of amino acids, vitamins or non essential amino acids can be added to a basal medium to enrich it and the resulting medium used as a classical MEM or BME. The final concentration of the MEM or BME must be 1X.