Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Septa
Replacement septa for VOA sampling vials
Replacement septa for VOA sampling vials
Replacement septa for VOA sampling vials
REST24694
:
Restek
Replacement septa for VOA sampling vials
REST24694
:
Restek
For:
VOA sampling vials, 60 ml sample collection vials, and 250 ml sample
Septum:
PTFE-lined silicone
Description:
Replacement septa, 24 mm×0,125"
Pk:
100