ContactUsLinkComponent
Sodium chloride 5 M in aqueous solution pH 8,0
Sodium chloride 5 M in aqueous solution pH 8,0
Catalog # J61807.AK
CAS Number:  
Sodium chloride 5 M in aqueous solution pH 8,0
Catalog # J61807.AK
CAS Number:  
Specifications
  • Pk:
    250 mL
  • Seq:
    2590
Specifications
About this item
 :
 :58.44 g/mol
Storage Temperature:Ambient
CAS Number:7647-14-5
Formula:NaCl
MDL Number:MFCD00003477