ContactUsLinkComponent
ESD boot covers, MICROGARD® 2000, model 407
ESD boot covers, MICROGARD® 2000, model 407
Catalog # ANSEWH20B-00407-00
Supplier:  Ansell
undefined
ESD boot covers, MICROGARD® 2000, model 407
Catalog # ANSEWH20B-00407-00
Supplier:  Ansell
Specifications
  • Shoe size:
    42 - 46
  • Garment application:
    Pharmaceutical,
    Electronics/semi-conductor
  • Colour:
    White
  • Pk:
    150 PAIR
Specifications
About this item
These ESD overboots are made from a microporous polyethylene film with a non woven inner layer.
  • Tie fastening
  • Elastic to top of boot
  • Bound seams
  • ESD PVC sole
  • Fit shoe size 42 - 46