ContactUsLinkComponent
Copper tubing for GC systems
Copper tubing for GC systems
  REST22628
 :  Restek
undefined
Copper tubing for GC systems
  REST22628
 :  Restek
  • Tubing ext.Ø:
    3,175 mm
  • Tubing int.Ø:
    1,651 mm
  • Wall thickness:
    0,07 mm
  • Pk:
    50 Feet
Copper tubing is compatible for use with a number of plumbing gas chromatography systems.
  • Use for plumbing GC systems
  • Cleaned to remove residual organics
  • Different inside and outside diameters available
Durable copper construction that can support a maximum pressure of 2,800 psig.