Orders LinkContactUsLinkComponent
Cryogenic vials, Nalgene®
Cryogenic vials, Nalgene®
Cryogenic vials, Nalgene®
Cryogenic vials, Nalgene®
Cryogenic vials, Nalgene®
These storage tubes are ideally suited for research and general laboratory storage of high-value and genomic materials down to the vapour phase of liquid nitrogen (at temperatures as low as –150 °C). They are made from PPCO and have HDPE screw caps.
  • External thread
  • Self standing
  • Non cytotoxic raw materials for added sample security and tubes certified free of Dnase and RNase for genomic applications
  • Tubes with writing area are graduated
System 100™ tubes allow for maximum storage capacity and have a PP closure and internal silicone gasket.
PS colour coders fit into the top of Nalgene® cryogenic vial closures (except Cat. No. 479-0006). They provide quick identification.
Warning: Do not store in the liquid phase of nitrogen without CryoFlex™ tubing.
PS colour coders fit into the top of Nalgene® cryogenic vial closures (except Cat. No. 479-0006). They provide quick identification.
Certifications: CE Marked for storage of diagnostic samples in the EU. Certified SAL of 10⁻⁶ for the highest sterility assurance available.
Caution: Do not store in the liquid phase of nitrogen without CryoFlex™ tubing.
  • Catalog No:
  • 479-3224
  • 479-3225
  • 479-3221
  • 479-0009
  • 479-0007
  • 479-3222
  • 479-0010
  • 479-0008
  • 479-3223
  • 479-0006
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
Product Family Options
System 100™ tubes
  • Description
    Capacity
    Base
    Ø×H
    Cap included
    Cap colour
  • System 100™ tubes, with writing area, sterile, RNase-, DNase-free
    1,00 ml
    Conical
    11,7×38,1 mm
    Yes
    Clear
    1 * 500 items
    1 * 500 items - 498,00 €
    498,00 €
    1 * 500 items
       498,00 €
      Low Manufacturing Impact
    Specifications
    Cat. No.
    479-3224
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • System 100™ tubes, with writing area, sterile, RNase-, DNase-free
    1,50 ml
    Conical
    11,7×47,5 mm
    Yes
    Clear
    1 * 500 items
    1 * 500 items - 615,00 €
    615,00 €
    1 * 500 items
       615,00 €
      Low Manufacturing Impact
    Specifications
    Cat. No.
    479-3225
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
Standard tubes
  • Description
    Capacity
    Base
    Ø×H
    Cap included
    Cap colour
  • Tubes, with writing area, sterile, RNase-, DNase-free
    1,20 ml
    Conical
    12,7×38,1 mm
    Yes
    Clear
    1 * 500 items
    1 * 500 items - 397,00 €
    397,00 €
    1 * 500 items
       397,00 €
      Low Manufacturing Impact
    Specifications
    Cat. No.
    479-3221
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • Tubes, with writing area, sterile, RNase-, DNase-free
    1,20 ml
    Conical
    12,7×38,1 mm
    Yes
    Clear
    1 * 1.000 items
    1 * 1.000 items - 745,00 €
    745,00 €
    1 * 1.000 items
       745,00 €
      Low Manufacturing Impact
    Specifications
    Cat. No.
    479-0009
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • Tubes, without writing area, non sterile
    1,20 ml
    Conical
    12,7×38,1 mm
    Yes
    Clear
    1 * 1.000 items
    1 * 1.000 items - 584,00 €
    584,00 €
    1 * 1.000 items
       584,00 €
      Low Manufacturing Impact
    Specifications
    Cat. No.
    479-0007
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • Tubes, with writing area, sterile, RNase-, DNase-free
    2,00 ml
    Conical
    12,7×47,5 mm
    Yes
    Clear
    1 * 500 items
    1 * 500 items - 400,00 €
    400,00 €
    1 * 500 items
       400,00 €
      Low Manufacturing Impact
    Specifications
    Cat. No.
    479-3222
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • Tubes, with writing area, sterile, RNase-, DNase-free
    2,00 ml
    Conical
    12,7×47,5 mm
    Yes
    Clear
    1 * 1.000 items
    1 * 1.000 items - 745,00 €
    745,00 €
    1 * 1.000 items
       745,00 €
      Low Manufacturing Impact
    Specifications
    Cat. No.
    479-0010
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • Tubes, without writing area, non sterile
    2,00 ml
    Conical
    12,7×47,5 mm
    Yes
    Clear
    1 * 1.000 items
    1 * 1.000 items - 584,00 €
    584,00 €
    1 * 1.000 items
       584,00 €
      Low Manufacturing Impact
    Specifications
    Cat. No.
    479-0008
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • Tubes, with writing area, sterile, RNase-, DNase-free
    5,00 ml
    Conical
    12,7×88,9 mm
    Yes
    Clear
    1 * 250 items
    1 * 250 items - 253,00 €
    253,00 €
    1 * 250 items
       253,00 €
      Low Manufacturing Impact
    Specifications
    Cat. No.
    479-3223
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • Tubes, without writing area, sterile, suitable for storage down to -80 °C only
    15,00 ml
    Flat
    33,3×46,0 mm
    Yes
    Clear
    1 * 75 items
    1 * 75 items - 188,00 €
    188,00 €
    1 * 75 items
       188,00 €
      Low Manufacturing Impact
    Specifications
    Cat. No.
    479-0006
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
Vial closure colour coders
  •  
    Colour
    Description
  • Red
    Colour coder
    1 * 100 items
    1 * 100 items - 19,50 €
    19,50 €
    1 * 100 items
       19,50 €
  •  
    Colour
    Description
  • White
    Colour coder
    1 * 100 items
    1 * 100 items - 19,50 €
    19,50 €
    1 * 100 items
       19,50 €
  •  
    Colour
    Description
  • Yellow
    Colour coder
    1 * 100 items
    1 * 100 items - 19,50 €
    19,50 €
    1 * 100 items
       19,50 €
  •  
    Colour
    Description
  • Blue
    Colour coder
    1 * 100 items
    1 * 100 items - 19,50 €
    19,50 €
    1 * 100 items
       19,50 €
  •  
    Colour
    Description
  • Green
    Colour coder
    1 * 100 items
    1 * 100 items - 19,50 €
    19,50 €
    1 * 100 items
       19,50 €
CryoSleeve™
  •  
    Description
  • Storage tube protection, 273 mm
    1 * 100 items
    1 * 100 items - 179,00 €
    179,00 €
    1 * 100 items
       179,00 €