Orders LinkContactUsLinkComponent
Mini regulator
Mini regulator
Mini gas regulator, with 0 - 15 psig outlet pressure gauge
Catalog # REST22032
Supplier:   Restek
Mini regulator
Catalog # REST22032
Supplier:  Restek
Mini regulator
Mini regulator
Mini gas regulator, with 0 - 15 psig outlet pressure gauge
Price
571,91 €
Log in for availability
1 * 1 items
Product Details & Documents
This mini regulator is suitable for natural gas and refinery gas standards.
Inlet pressure range: 0 to 20,68 bar
Outlet pressure range: 0 to 1,03 bar
Delivery information: Supplied with outlet pressure gauge (0 - 1,03 bar), brass CGA 170 nut and nipple, and compression outlet (3,175 mm).
Specifications
  • Description:
    Mini gas regulator, with 0 - 15 psig outlet pressure gauge