Thermal transfer printer labels, type B-351, Defender
Printer labels, B-351, THT-17-7351-3, Vinyl, Permanent acrylic, Thermal transfer, White, 360X-Plus II, 50×25 mm, 3000 labels/roll
BRDY020822
: Brady
Printable, tamper-evident vinyl labels for thermal transfer printers.
- Fractures easily
- Prevents one-piece removal
Labels are designed to fracture easily in order to show signs of product tampering and to prevent one-piece label removal. Use caution when removing from liner as material is fragile.
Documents
TDS B-7351 Labels and Signs
Datasheet
- Printer model:360X-Plus II
- Printing method:Thermal transfer
- Type:B-351
- Colour:White
- Description:Printer labels
- Dimensions:50×25 mm
- Material:Vinyl
- Model:THT-17-7351-3
- Packed:3000 labels/roll
- Adhesive:Permanent acrylic