ContactUsLinkComponent
Guard columns, Metal MXT®, Restek
  REST70067
 :  Restek
undefined
Guard columns, Metal MXT®, Restek
  REST70067
 :  Restek
  • Column Format:
    Guard column
  • Ø int.:
    0,53 mm
  • Length:
    5 m
  • Phase:
    MXT®-Polar Deactivated
  • Pk:
    1
These columns are made up of passivated stainless steel.
  • Polyethylene glycol deactivation layer provides optimum wettability for polar compounds
  • Minimise peak splitting when using polar solvents such as methanol or water
  • Maximum temperature: 280 °C