Orders LinkContactUsLinkComponent
PLOT columns, MXT® metal
PLOT columns, MXT® metal
Supplier:   Restek
PLOT columns, MXT® metal
Supplier:  Restek
Certificates
PLOT columns, MXT® metal
PLOT columns, MXT® metal
11 Options Available Below
Product Details & Documents
PLOT columns are made from Siltek® treated stainless steel and have the same characteristics and performance as fused silica PLOT columns but useful for GCs and other field applications.
  • Made into small coil diameters and perfect for tight spaces
  • Rugged material withstands rough handling and shock
  • Available in 3,5" coil diameter or 7" diameter with 11-pin cage
Product Family Options
MXT®-Msieve 5A
Product Information
  • Column Format
    Phase
    Film thickness
    Ø int.
    Length
    Availability
    Price
  • 3,5" coil
    Capillary
    MXT®-Msieve 5A
    20,00 µm
    0,25 mm
    15 m
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items
  • 3,5" coil
    Capillary
    MXT®-Msieve 5A
    50,00 µm
    0,53 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 1 783,00 €
    1 783,00 €
    1 * 1 items
       1 783,00 €
  • REST79723
    7" coil, 11-pin cage
    7" coil, 11-pin cage
    Capillary
    MXT®-Msieve 5A
    50,00 µm
    0,53 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 1 783,00 €
    1 783,00 €
    1 * 1 items
       1 783,00 €
MXT®-Alumina BOND/Na2SO4
Product Information
  • Column Format
    Phase
    Film thickness
    Ø int.
    Length
    Availability
    Price
  • REST79714
    7" coil, 11-pin cage
    7" coil, 11-pin cage
    Capillary
    MXT®-Alumina BOND/Na2SO4
    10,00 µm
    0,53 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 1 166,00 €
    1 166,00 €
    1 * 1 items
       1 166,00 €
  • 3,5" coil
    Capillary
    MXT®-Alumina BOND/Na2SO4
    10,00 µm
    0,53 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 1 166,00 €
    1 166,00 €
    1 * 1 items
       1 166,00 €
MXT®-Q-BOND
Product Information
  • Column Format
    Phase
    Film thickness
    Ø int.
    Length
    Availability
    Price
  • REST79718
    7" coil, 11-pin cage
    7" coil, 11-pin cage
    Capillary
    MXT®-Q-BOND
    8,00 µm
    0,25 mm
    15 m
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 890,00 €
    890,00 €
    1 * 1 items
       890,00 €
  • 3,5" coil
    Capillary
    MXT®-Q-BOND
    8,00 µm
    0,25 mm
    15 m
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 890,00 €
    890,00 €
    1 * 1 items
       890,00 €
  • REST79716
    7" coil, 11-pin cage
    7" coil, 11-pin cage
    Capillary
    MXT®-Q-BOND
    20,00 µm
    0,53 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 1 243,00 €
    1 243,00 €
    1 * 1 items
       1 243,00 €
  • 3,5" coil
    Capillary
    MXT®-Q-BOND
    20,00 µm
    0,53 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 1 243,00 €
    1 243,00 €
    1 * 1 items
       1 243,00 €
MXT®-S-BOND
Product Information
  • Column Format
    Phase
    Film thickness
    Ø int.
    Length
    Availability
    Price
  • 3,5" coil
    Capillary
    MXT®-S-BOND
    20,00 µm
    0,53 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items
  • REST79712
    7" coil, 11-pin cage
    7" coil, 11-pin cage
    Capillary
    MXT®-S-BOND
    20,00 µm
    0,53 mm
    30 m