MMP inhibitor profiling kit
Catalog # ENZOBMLAK3080001
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Pk:1 KIT
- Description:MMP inhibitor profiling kit, fluorometric, red
- Seq:02
Specifications
About this item
The MMP inhibitor profiling kit, fluorometric RED is a complete assay system designed to examine the specificity of inhibitors against a panel of ten matrix metalloproteinase enzymes, using a quenched fluorogenic substrate OMNIMMP® RED.
The assays are performed in a convenient 96-well microplate format. The compound NNGH is also included as a prototypic control inhibitor.