Human CRP (C Reactive Protein) ELISA kit
Catalog # BIRBORB78731-96
Supplier: Biorbyt
CAS Number:
Specifications
- Pk:96
- Primary antibody reactivity:Human
- Description:Human CRP (C Reactive Protein) ELISA Kit
- Detection range:2 - 600 pg/ml
- Sensitivity:2 pg/ml
Specifications
About this item
Human CRP (C Reactive Protein) ELISA kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human CRP in serum, plasma, cell culture supernatants and urine.
Delivery: Kit includes precoated 96-well microplate, wash buffer concentrate 20X, 25 ml; standards, 2 vials; assay diluent D 5X, 2×15 ml; assay diluent B 5X, 15 ml; detection antibody CRP, 2 vials; HRP-streptavidin concentrate 300X, 200 µl; TMB one-step substrate reagent, 12 ml; and stop solution, 8 ml.