ContactUsLinkComponent
Detergents for glassware washers, ProCare Lab 11 AP
Detergents for glassware washers, ProCare Lab 11 AP
Catalog # MIEL10266540
Supplier:  MIELE
undefined
Detergents for glassware washers, ProCare Lab 11 AP
Catalog # MIEL10266540
Supplier:  MIELE
Specifications
  • Capacity:
    10 kg
  • Description:
    Alkaline detergent powder, ProCare Lab 11 AP
  • Pk:
    10 kg
  • pH range:
    11,5 - 11,7
Specifications
About this item
Powder, alkaline detergent for machine based cleaning of laboratory glassware and laboratory utensils.
  • Removes inorganic and organic soils
  • Free from phosphates
  • Can be used in the water hardness range up to 6°dH
ProCare Lab 11 AP is suitable for general purpose use in water laboratories, serological institutes and in laboratories in the phosphate industry.
Note: Suitable for stainless steel, laboratory glassware, plastics such as PE, PP, PVDF, PTFE. Not suitable for use on non ferrous metals, light alloys, anodised aluminium, and aluminium.