Antimicrobial hand cleansers, HiBiScrub®
Supplier: MOLNLYCKE HEALTH CARE
Certificates
About this item
HiBiScrub® is an antimicrobial skin cleanser containing 4% Chlorhexidine gluconate. It binds to the skin forming an effective barrier that will keep killing bacteria and protecting long after application, showing up to 6 hours residual effect and increased efficacy upon repeated application.
- Effective kill rate for many organisms that are implicated with healthcare associated infections within one minute
- Active chlorhexidine gluconate is extremely effective against a wide range of Gram positive and Gram negative bacteria, yeast, fungi and viruses
- Contains a blend of emollients, dermatologically tested HiBiScrub helps soften and hydrate the skin
- Proven to be significantly less irritating than a leading 4% Chlorhexidine gluconate solution
- Reduces moisture loss even with frequent use
- Short and effective contact times and proven short contact times on listed test standards
Frequently Bought Together