Capillary columns, Rtx®-VMS (fused silica)
: Restek
Proprietary Crossbond® phase, specific columns for volatile organic compounds.
- Complete separation of U.S. EPA Method 8260B compounds in less than 10 minutes
- Offers lower bleed, better selectivity, and overall faster analysis
- Stable to 260 °C
0,18 and 0,25 mm ID columns allow sample splitting at the injection port, eliminating the added expense and maintenance of a jet separator. 0,45 mm or 0,53 mm ID column can be directly connected to the purge-and-trap transfer line in a system equipped with a jet separator.
Frequently Bought Together