ContactUsLinkComponent
Microplate Clamp for VWR® Standard and Advanced Shakers
Microplate Clamp for VWR® Standard and Advanced Shakers
Catalog # 444-2954
Supplier:  VWR Collection
undefined
Microplate Clamp for VWR® Standard and Advanced Shakers
Catalog # 444-2954
Supplier:  VWR Collection
Specifications
  • For:
    Standard/Advanced 3500, 5000, 10000, 15000 shakers
  • Description:
    Microplate clamp, holds 1 standard microplate or 1 deep-well plate, 89×129 mm
  • Seq:
    0170
  • Environmentally Preferable:
  • Pk:
    1
Specifications
About this item
Attaches to the relevant universal platform being used with the shaker. For details of universal platforms please see the relevant entries.