Storage plates and blocks, 96-well, Corning®
Catalog # CORN3947
Supplier: Corning
Specifications
- Packed:5/bag
- Sterile:-
- Description:Deep well plate, 96-well, natural, V-bottom square well
- Seq:03
- W×D×H:127,76×85,48×44,2 mm
- Environmentally Preferable:
- Pk:50
- Well volume:2200 µl
Specifications
About this item
PP, non treated, without lid. Standard and large volume 96-well microplates and storage blocks to meet assay and storage requirements.
- Choice of flat, round or V-shaped well bottom
- Uniform skirt heights for greater robotic gripping surface
- Resistant to many common organic solvents (e.g. DMSO, ethanol, methanol)
- Certified free from DNase and RNase