Orders LinkContactUsLinkComponent
Screw thread vials, 8-425
Screw thread vials, 8-425
Supplier:   Restek
Screw thread vials, 8-425
Supplier:  Restek
Certificates
Screw thread vials, 8-425
Screw thread vials, 8-425
3 Options Available Below
Product Details & Documents
Screw thread vials fits all 2,0 ml, 12×32 mm vial based autosamplers and provide most secure environment for your sample because evaporation is minimised.
Product Family Options
Product Information
  • Capacity
    Ø×H
    Colour
    Cap included
    Cap Size
    Availability
    Price
  • REST24619
    Screw thread vial
    Screw thread vial
    2,00 ml
    12×32 mm
    Clear
    No
    8-425
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items - 39,00 €
    39,00 €
    1 * 100 items
       39,00 €
  • REST24620
    Screw thread vial
    Screw thread vial
    2,00 ml
    12×32 mm
    Clear
    No
    8-425
    Log in for availability
    Log in for availability
    1 * 1.000 items
    1 * 1.000 items - 313,00 €
    313,00 €
    1 * 1.000 items
       313,00 €
  • REST24622
    Screw thread vial
    Screw thread vial
    2,00 ml
    12×32 mm
    Amber
    No
    8-425
    Log in for availability
    Log in for availability
    1 * 1.000 items
    1 * 1.000 items - 358,00 €
    358,00 €
    1 * 1.000 items
       358,00 €
Caps
Product Information
  • Catalog #
    Description
    Availability
    Price
  • Open-hole caps, PP, black
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items - 14,00 €
    14,00 €
    1 * 100 items
       14,00 €
Product Information
  • Catalog #
    Description
    Availability
    Price
  • Open-hole caps, PP, black
    Log in for availability
    Log in for availability
    1 * 1.000 items
    1 * 1.000 items - 92,00 €
    92,00 €
    1 * 1.000 items
       92,00 €
Septa
Product Information
  • Catalog #
    Ø
    Type
    Thickness
    Availability
    Price
  • 8 mm
    Red PTFE/white silicone
    1,65 mm
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items - 37,00 €
    37,00 €
    1 * 100 items
       37,00 €
Product Information
  • Catalog #
    Ø
    Type
    Thickness
    Availability
    Price
  • 8 mm
    Red PTFE/silicone
    1,52 mm
    Log in for availability
    Log in for availability
    1 * 1.000 items
    1 * 1.000 items - 232,00 €
    232,00 €
    1 * 1.000 items
       232,00 €
Product Information
  • Catalog #
    Ø
    Type
    Thickness
    Availability
    Price
  • 8 mm
    Red PTFE/white silicone
    1,65 mm
    Log in for availability
    Log in for availability
    1 * 1.000 items
    1 * 1.000 items - 256,00 €
    256,00 €
    1 * 1.000 items
       256,00 €
Product Information
  • Catalog #
    Ø
    Type
    Thickness
    Availability
    Price
  • 8 mm
    Red PTFE/silicone
    1,52 mm
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items - 35,00 €
    35,00 €
    1 * 100 items
       35,00 €
Caps and septa (preassembled)
Product Information
  • Catalog #
    Thread
    Septum
    Ø
    Description
    Availability
    Price
  • 8-425
    Red PTFE/silicone, 1,65 mm
    8 mm
    Caps and septa, preassembled
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items - 46,00 €
    46,00 €
    1 * 100 items
       46,00 €
Product Information
  • Catalog #
    Thread
    Septum
    Ø
    Description
    Availability
    Price
  • 8-425
    PTFE/silicone
    8 mm
    Caps and septa, preassembled, top hat
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items - 60,00 €
    60,00 €
    1 * 100 items
       60,00 €
Product Information
  • Catalog #
    Thread
    Septum
    Ø
    Description
    Availability
    Price
  • 8-425
    Red PTFE/silicone, 1,65 mm
    8 mm
    Caps and septa, preassembled
    Log in for availability
    Log in for availability
    1 * 1.000 items
    1 * 1.000 items - 389,00 €
    389,00 €
    1 * 1.000 items
       389,00 €
Product Information
  • Catalog #
    Thread
    Septum
    Ø
    Description
    Availability
    Price
  • 8-425
    PTFE/silicone
    8 mm
    Caps and septa, preassembled, top hat
    Log in for availability
    Log in for availability
    1 * 1.000 items
    1 * 1.000 items - 507,00 €
    507,00 €
    1 * 1.000 items
       507,00 €