Orders LinkContactUsLinkComponent
ALLN
ALLN
10 mg
Catalog # 786-057
Supplier:   G-Biosciences
ALLN
Catalog # 786-057
Supplier:  G-Biosciences
CAS Number:  
CAS Number:  
Price
304,00 €
Log in for availability
1 * 10 mg
Product Details & Documents
ALLN is a cell permeable peptide aldehyde inhibitor of calpain I and to a lesser extent calpain II. It also inhibits other neutral cysteine proteases, cathepsin B and L and the proteasome.
Specifications
Specifications
  • Pk:
    10 mg
  • Cat. No.:
    786-057
  • Protein/peptide name:
    ALLN
Specifications